2-Bromo-1-chloro-3-fluorobenzene 98%

Supplier: Ambeed
Warning

A260119-100G A260119-10G A260119-1G A260119-25G A260119-5G
103812-478EA 11 USD
103812-478 103852-360 76786-984 76786-986 77091-936 77091-938 77091-940
2-Bromo-1-chloro-3-fluorobenzene 98%
2-Bromo-1-chloro-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClF
Boiling Pt: 198 °C (760 mm Hg)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD09878161
CAS Number: 309721-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR