2-Bromo-4'-(trifluoromethyl)acetophenone 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one

A153285-100G A153285-1G A153285-500G A153285-25G A153285-5G A153285-10G
103842-662EA 18 USD
103842-662 76768-226 76768-228 76768-230 77318-312 77318-314 77549-926
2-Bromo-4'-(trifluoromethyl)acetophenone 97%
2-Bromo-4'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O
MW: 267.0400 g/mol
Boiling Pt: 264.2 °C (760 mm Hg)
Melting Pt: 45…46 °C
Flash Pt: 113.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00126489
CAS Number: 383-53-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR