2-Bromo-1,4-dimethoxybenzene 98%

Supplier: Ambeed
Warning

A105975-100G A105975-10G A105975-25G A105975-500G A105975-5G
103822-726EA 7 USD
103822-726 77093-746 77093-748 77093-750 77093-752 77309-586
2-Bromo-1,4-dimethoxybenzene 98%
2-Bromo-1,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
Boiling Pt: 269.6 °C (760 mm Hg)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00008355
CAS Number: 25245-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR