2-Bromo-1,4-dichlorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,5-dichlorobenzene

A205869-25G A205869-100G A205869-10G A205869-500G A205869-5G
103848-336EA 5 USD
103848-336 77086-566 77152-314 77152-316 77152-318 77302-972
2-Bromo-1,4-dichlorobenzene 98%
2-Bromo-1,4-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 236.2 °C (760 mm Hg)
Melting Pt: 32…36 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00018505
CAS Number: 1435-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR