2-Bromo-1,3-dimethoxybenzene 98%

Supplier: Ambeed
Warning

A371663-5G A371663-100G A371663-10G A371663-500G A371663-1G A371663-25G
103826-486EA 9 USD
103826-486 103837-504 76840-756 77240-254 77240-256 77240-258 77306-832 77306-834
2-Bromo-1,3-dimethoxybenzene 98%
2-Bromo-1,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 250 °C (760 mm Hg)
Melting Pt: 91…94 °C
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD07780177
CAS Number: 16932-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR