2-Bromo-1,3-diisopropylbenzene 96%

Supplier: Ambeed
Warning

A143129-10G A143129-25G A143129-1G A143129-5G A143129-100G
103803-240EA 15 USD
103803-240 76871-302 77299-326 77316-486 77316-488 77592-696
2-Bromo-1,3-diisopropylbenzene 96%
2-Bromo-1,3-diisopropylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇Br
MW: 241.17 g/mol
Boiling Pt: 254.3±9.0 °C (760 mm Hg)
Flash Pt: 104.1 °C
Storage Temperature: Ambient
MDL Number: MFCD16876782
CAS Number: 57190-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR