2-Bromo-1,3-diethyl-5-methylbenzene 97%
Supplier: Ambeed
|
Warning
|
A123844-100G
A123844-10G
A123844-25G
A123844-5G
A123844-1G
103800-450EA
13
USD
103800-450
76822-590
76822-592
76822-594
77297-960
77312-570
2-Bromo-1,3-diethyl-5-methylbenzene 97%
2-Bromo-1,3-diethyl-5-methylbenzene
Precaución: for Research Use Only
Formula:
C₁₁H₁₅Br MW: 227.14 g/mol Boiling Pt: 259 °C (760 mm Hg) Melting Pt: 2 °C Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD16036168 CAS Number: 314084-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...