4-Bromo-3,5-dichlorofluorobenzene 98%

Supplier: Ambeed
Warning

A333504-1G A333504-25G A333504-5G A333504-250MG
103825-648EA 7 USD
103825-648 103836-614 76839-616 76839-618 76839-620 77032-944
4-Bromo-3,5-dichlorofluorobenzene 98%
4-Bromo-3,5-dichlorofluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
Storage Temperature: Ambient
MDL Number: MFCD11845890
CAS Number: 263333-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR