1-Bromo-2,6-bis(trifluoromethyl)benzene 98%
Supplier: Ambeed
|
Warning
|
A192229-1G
A192229-5G
A192229-100G
A192229-10G
A192229-250MG
103816-426EA
15
USD
103816-426
103847-032
77084-906
77084-908
77151-918
77151-920
77151-922
1-Bromo-2,6-bis(trifluoromethyl)benzene 98%
1-Bromo-2,6-bis(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₈H₃BrF₆ Boiling Pt: 159 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00236210 CAS Number: 118527-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...