1-Bromo-2,4,6-tri-tert-butylbenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1,3,5-tri-tert-butylbenzene

A348258-1G A348258-100G A348258-100MG A348258-250MG
103825-990EA 8 USD
103825-990 103836-978 76840-056 77239-962 77239-964 77239-966
1-Bromo-2,4,6-tri-tert-butylbenzene 97%
1-Bromo-2,4,6-tri-tert-butylbenzene

Precaución: for Research Use Only

Formula: C₁₈H₂₉Br
MW: 325.3300 g/mol
Boiling Pt: 299.7 °C (760 mm Hg)
Melting Pt: 168…173 °C
Flash Pt: 123.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00191861
CAS Number: 3975-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR