2-Bromo-1,3,5-triisopropylbenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,4,6-triisopropylbenzene

A153730-100G A153730-10G A153730-500G A153730-25G
103842-734EA 5 USD
103842-734 77235-544 77235-546 77235-548 77300-026
2-Bromo-1,3,5-triisopropylbenzene 97%
2-Bromo-1,3,5-triisopropylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₂₃Br
MW: 283.25 g/mol
Boiling Pt: 296.5 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051547
CAS Number: 21524-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR