2-Bromo-1,1-dimethoxypropane 95%

Supplier: Ambeed
Warning

A256383-10G A256383-250MG A256383-1G A256383-5G
103812-264EA 12 USD
103812-264 103852-128 77031-702 77031-704 77091-624 77304-674
2-Bromo-1,1-dimethoxypropane 95%
2-Bromo-1,1-dimethoxypropane

Precaución: for Research Use Only

Formula: C₅H₁₁BrO₂
Boiling Pt: 166.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11505877
CAS Number: 33170-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR