Bromoacetaldehyde dimethyl acetal 98% stabilised

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1,1-dimethoxyethane

Stabilization: 0.5% Stabilised with potassium carbonate

A297732-10G A297732-25G
103809-998EA 90 USD
103809-998 76816-694 76837-976
Bromoacetaldehyde dimethyl acetal 98% stabilised
Bromoacetaldehyde dimethyl acetal

Precaución: for Research Use Only

Formula: BrH₂CCH(OCH₃)₂
MW: 169.02 g/mol
Boiling Pt: 148
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000213
CAS Number: 7252-83-7
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR