2-Bromo-1,10-phenanthroline 97%

Supplier: Ambeed
Warning

A188184-1G A188184-5G A188184-10G A188184-250MG A188184-25G A188184-100G
103816-078EA 8 USD
103816-078 103846-668 77084-396 77084-398 77260-444 77260-446 77260-448 77746-208
2-Bromo-1,10-phenanthroline 97%
2-Bromo-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₂H₇BrN₂
MW: 259.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276287
CAS Number: 22426-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR