ω-Bromo-4-methylacetophenone 98%

Supplier: Ambeed
Danger

Synonyms: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

A459097-100G A459097-10G A459097-1G A459097-25G A459097-5G
103827-892EA 9 USD
103827-892 103838-968 76818-404 76818-406 77307-414 77307-416 77307-418
ω-Bromo-4-methylacetophenone 98%
ω-Bromo-4-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.07 g/mol
Boiling Pt: 263.6 °C (760 mm Hg)
Melting Pt: 45…49 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000203
CAS Number: 619-41-0
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR