2-Bromo-4'-(trifluoromethoxy)acetophenone 97%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone, 4-(Trifluoromethoxy)phenacyl bromide

A119809-100G A119809-250MG A119809-25G A119809-1G A119809-5G A119809-10G
103799-844EA 6 USD
103799-844 77258-052 77258-054 77258-056 77311-950 77311-952 77571-194
2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₂
MW: 283.04 g/mol
Melting Pt: 48…55 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052333
CAS Number: 103962-10-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR