2-Bromo-4'-(dimethylamino)acetophenone 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Dimethylamino)phenacyl bromide, 2-Bromo-1-(4-(dimethylamino)phenyl)ethanone

A184854-1G A184854-250MG A184854-25G
103815-800EA 93 USD
103815-800 103846-366 77083-994 77260-354 77301-930
2-Bromo-4'-(dimethylamino)acetophenone 95%
2-Bromo-4'-(dimethylamino)acetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
MW: 242.1124 g/mol
Boiling Pt: 330.2 °C (760 mm Hg)
Melting Pt: 89…92 °C
Flash Pt: 153.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06409221
CAS Number: 37904-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR