3-Bromo-2-fluorophenacyl bromide 95%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1-(3-bromo-2-fluorophenyl)ethan-1-one, 2,3'-Dibromo-2'-fluoroacetophenone, 2-Bromo-1-(3-bromo-2-fluorophenyl)ethanone

A119677-100MG A119677-250MG A119677-1G
103799-816EA 128 USD
103799-816 77189-602 77189-604 77297-712
3-Bromo-2-fluorophenacyl bromide 95%
3-Bromo-2-fluorophenacyl bromide

Precaución: for Research Use Only

Formula: C₈H₅Br₂FO
MW: 295.931 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16140216
CAS Number: 1204333-47-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR