2-Bromo-1-(3,4-dimethoxyphenyl)ethanone 97%

Supplier: Ambeed
Danger

Synonyms: 3,4-Dimethoxyphenacyl bromide

A205596-100G A205596-10G A205596-25G A205596-1G
103817-646EA 16 USD
103817-646 103848-312 76647-492 76647-494 76647-496 77086-536
2-Bromo-1-(3,4-dimethoxyphenyl)ethanone 97%
2-Bromo-1-(3,4-dimethoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MW: 259.11 g/mol
Boiling Pt: 326.5 °C (760 mm Hg)
Melting Pt: 81…83 °C
Flash Pt: 151.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425186
CAS Number: 1835-02-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR