ω-Bromo-2-methoxyacetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methoxyphenacyl bromide, α-Bromo-2-methoxyacetophenone, 2-Bromo-2'-methoxyacetophenone, 2-Bromo-1-(2-methoxyphenyl)ethanone, 2-Bromo-1-(2-methoxyphenyl)ethan-1-one, Bromomethyl o-methoxyphenyl ketone

A475501-10G A475501-1G A475501-25G A475501-5G
103828-072EA 13 USD
103828-072 103839-144 76695-452 76843-072 76843-074 77307-516
ω-Bromo-2-methoxyacetophenone 98%
ω-Bromo-2-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 324
Melting Pt: 43…45 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000196
CAS Number: 31949-21-0
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR