2-Bromo-1-(2-bromoethyl)-4-chlorobenzene 95%

Supplier: Ambeed
Warning

A800792-5G A800792-250MG A800792-25G
103841-064EA 94 USD
103841-064 76845-306 76993-288 77886-787
2-Bromo-1-(2-bromoethyl)-4-chlorobenzene 95%
2-Bromo-1-(2-bromoethyl)-4-chlorobenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂Cl
Storage Temperature: Refrigerator
MDL Number: MFCD22376643
CAS Number: 52927-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR