2-Boc-aminothiazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-1,3-thiazole-4-carboxylic acid, 2-BOC protected, 2-Boc-Aminothiazole-4-carboxylic acid

A110127-10G A110127-250MG A110127-1G A110127-5G
103797-622EA 19 USD
103797-622 103823-430 77160-688 77160-690 77310-230 77310-232
2-Boc-aminothiazole-4-carboxylic acid 98%
2-Boc-aminothiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄S
MW: 244.2676 g/mol
Melting Pt: 250…252 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02181057
CAS Number: 83673-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR