2-Boc-6-amino-2-azaspiro[3.3]heptane 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2-azaspiro[3.3]heptane, N2-BOC protected, tert-Butyl 6-amino-2-azaspiro[3.3]heptane-2-carboxylate

A142212-100MG A142212-250MG A142212-1G A142212-25G
103803-124EA 57 USD
103803-124 76972-658 76972-660 77316-338 77671-030
2-Boc-6-amino-2-azaspiro[3.3]heptane 97%
2-Boc-6-amino-2-azaspiro[3.3]heptane

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MW: 212.2900 g/mol
Melting Pt: 115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15071432
CAS Number: 1211586-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR