Hexahydrocyclopenta[c]pyrrol-5(1H)-one, N-BOC protected 95%

Supplier: Ambeed
Warning

Synonyms: 2-(tert-Butoxycarbonyl)-5-oxooctahydrocyclopenta[c]pyrrole, 2-Boc-5-oxohexahydrocyclopenta[c]pyrrole, tert-Butyl 5-oxooctahydrocyclopenta[c]pyrrole-2-carboxylate

A133732-10G A133732-250MG A133732-5G A133732-1G A133732-25G
103801-910EA 16 USD
103801-910 76811-370 76811-372 77298-656 77314-594 77314-596
Hexahydrocyclopenta[c]pyrrol-5(1H)-one, N-BOC protected 95%
Hexahydrocyclopenta[c]pyrrol-5(1H)-one, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₃
MW: 225.284 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11616063
CAS Number: 148404-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR