2-Boc-2-azabicyclo[2.2.1]hept-5-ene 97%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylate, 2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]hept-5-ene, 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylic acid tert-Butyl ester, 2-Boc-2-azabicyclo[2.2.1]hept-5-ene

A101562-1G A101562-5G A101562-25G A101562-10G
103796-028EA 24 USD
103796-028 103821-714 77297-152 77297-154 77599-480 77666-468
2-Boc-2-azabicyclo[2.2.1]hept-5-ene 97%
2-Boc-2-azabicyclo[2.2.1]hept-5-ene

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₂
Boiling Pt: 259.565 °C (760 mm Hg)
Flash Pt: 110.781 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22416354
CAS Number: 188345-71-3
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR