2-Benzylidene-4-methyl-3-oxo-N-phenylpentanamide 97%

Supplier: Ambeed
Warning

A108770-10G A108770-500G
103823-176EA 17 USD
103823-176 77257-562 77745-813
2-Benzylidene-4-methyl-3-oxo-N-phenylpentanamide 97%
2-Benzylidene-4-methyl-3-oxo-N-phenylpentanamide

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₂
MW: 293.36 g/mol
Boiling Pt: 519.438 °C (760 mm Hg)
Melting Pt: 143…154 °C
Flash Pt: 188.668 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671709
CAS Number: 125971-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR