2-Benzylacrylic acid 98%

Supplier: Ambeed
Warning

A113587-100G A113587-10G A113587-25G A113587-500G A113587-5G
103808-104EA 285 USD
103808-104 76971-036 76971-038 76971-040 76971-042 76971-044
2-Benzylacrylic acid 98%
2-Benzylacrylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
Boiling Pt: 326.1 °C (760 mm Hg)
Melting Pt: 66…68 °C
Flash Pt: 230.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00854174
CAS Number: 5669-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR