Clorofene 96%

Supplier: Ambeed
Danger

Synonyms: 2-Benzyl-4-chlorophenol

A335646-25G A335646-5G A335646-10G A335646-100G
103836-666EA 6 USD
103836-666 76839-676 76839-678 77003-056 77601-510
Clorofene 96%
Clorofene

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO
MW: 218.68 g/mol
Boiling Pt: 160…162 °C (760 mm Hg)
Melting Pt: 46…49 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00020140
CAS Number: 120-32-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR