2-Benzoxazolinone 98%

Supplier: Ambeed
Warning

Synonyms: benzo[d]oxazol-2(3H)-one

A171201-100G A171201-10G A171201-500G A171201-25G A171201-5G
103844-878EA 5 USD
103844-878 76813-200 76813-202 76813-204 77082-008 77301-106
2-Benzoxazolinone 98%
2-Benzoxazolinone

Precaución: for Research Use Only

Formula: C₇H₅NO₂
MW: 135.12 g/mol
Boiling Pt: 299 °C (760 mm Hg)
Melting Pt: 137…139 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00005716
CAS Number: 59-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR