2-Aminoterephthalic acid 95%

Supplier: Ambeed
Warning

A307249-100G A307249-1G A307249-500G A307249-25G A307249-5G
103835-672EA 10 USD
103835-672 76648-128 76648-130 76648-132 76838-404 76838-406
2-Aminoterephthalic acid 95%
2-Aminoterephthalic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
Melting Pt: 324 °C
Flash Pt: 226.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00134536
CAS Number: 10312-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR