2-Aminopyrimidine-5-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminopyrimidine-5-carboxaldehyde

A135535-100MG A135535-10G A135535-250MG A135535-1G A135535-5G
103802-174EA 12 USD
103802-174 77109-500 77109-502 77109-504 77315-010 77315-012
2-Aminopyrimidine-5-carbaldehyde 98%
2-Aminopyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₅N₃O
MW: 123.11 g/mol
Melting Pt: 209…214 °C
Flash Pt: 179.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03701714
CAS Number: 120747-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR