2-Aminopyrimidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A407883-100MG A407883-250MG
103827-166EA 103 USD
103827-166 103838-208 77264-556 77264-558
2-Aminopyrimidine-4-carboxylic acid 97%
2-Aminopyrimidine-4-carboxylic acid

Precaución: Keep in a dark place, sealed in dry, at 2…8°C

Formula: C₅H₅N₃O₂
Boiling Pt: 474.6 °C (760 mm Hg)
Flash Pt: 240.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07783936
CAS Number: 2164-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR