Isocytosine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4(1H)-pyrimidinone, 2-Aminopyrimidin-4(1H)-one

A166226-100G A166226-10G A166226-1G A166226-25G A166226-5G
103806-030EA 7 USD
103806-030 103844-290 76872-188 76872-190 77300-812 77300-814 77300-816
Isocytosine 97%
Isocytosine

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MW: 111.10 g/mol
Flash Pt: 106.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00057557
CAS Number: 108-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR