2-Aminopyridin-4-ol 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-hydroxypyridine

A121852-100G A121852-10G A121852-25G A121852-1G A121852-5G
103800-144EA 6 USD
103800-144 76971-568 76971-570 76971-572 77312-272 77312-274
2-Aminopyridin-4-ol 95%
2-Aminopyridin-4-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 253.9 °C (760 mm Hg)
Melting Pt: 130…134 °C
Flash Pt: 107.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114161
CAS Number: 33631-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR