2-Aminopropanenitrile hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A158091-1G
A158091-100MG
103805-092EA
272
USD
103805-092
77300-314
77883-243
2-Aminopropanenitrile hydrochloride 95%
2-Aminopropanenitrile hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₇ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09971651 CAS Number: 72187-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...