2-Amino-1,3-propanediol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol
A601557-25G
A601557-10G
103810-474EA
164
USD
103810-474
103840-120
76844-284
77170-116
2-Amino-1,3-propanediol 98%
2-Amino-1,3-propanediol
Precaución: for Research Use Only
Formula:
C₃H₉NO₂ MW: 91.11 g/mol Boiling Pt: 266.7 °C (760 mm Hg) Melting Pt: 52…55 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051487 CAS Number: 534-03-2 UN: 3259 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...