2-Amino-1,1,3-tricyano-1-propene 95%

Supplier: Ambeed
Warning

Synonyms: Malononitrile Dimer, 2-Aminoprop-1-ene-1,1,3-tricarbonitrile

A450287-5G A450287-10G
103838-858EA 24 USD
103838-858 76842-674 77252-772
2-Amino-1,1,3-tricyano-1-propene 95%
2-Amino-1,1,3-tricyano-1-propene

Precaución: for Research Use Only

Formula: C₆H₄N₄
MW: 132.13 g/mol
Boiling Pt: 591.9 °C (760 mm Hg)
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00001851
CAS Number: 868-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR