2-Aminophenylboronic acid hydrochloride 97%

Supplier: Ambeed
Warning

A229349-1G A229349-25G A229349-5G A229349-10G A229349-250MG
103819-470EA 14 USD
103819-470 103850-370 77089-208 77089-210 77089-212 77112-930 77112-932
2-Aminophenylboronic acid hydrochloride 97%
2-Aminophenylboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉BClNO₂
MW: 173.4052 g/mol
Melting Pt: 145…156 °C
Flash Pt: 163.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02258096
CAS Number: 863753-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR