2-Amino-N-methylbenzamide 98%

Supplier: Ambeed
Warning

A139144-100G A139144-10G A139144-25G
77190-324EA 170.98 USD
77190-324 77190-326 77299-046
2-Amino-N-methylbenzamide 98%
2-Amino-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
Boiling Pt: 346.7 °C (760 mm Hg)
Melting Pt: 75 °C
Flash Pt: 163.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00060602
CAS Number: 4141-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR