2-Amino-N-methylacetamide 97%

Supplier: Ambeed
Warning

A197320-250MG A197320-1G A197320-5G
103816-910EA 25 USD
103816-910 103847-532 76873-266 76873-268 77085-530 77085-532
2-Amino-N-methylacetamide 97%
2-Amino-N-methylacetamide

Precaución: for Research Use Only

Formula: C₃H₈N₂O
Boiling Pt: 275.6 °C (760 mm Hg)
Flash Pt: 120.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06375945
CAS Number: 22356-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR