2-Aminonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid

A321141-25G A321141-100G A321141-10G A321141-5G A321141-500G
103836-198EA 9 USD
103836-198 76839-092 77277-704 77277-706 77306-234 77593-904
2-Aminonicotinic acid 97%
2-Aminonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 342.3 °C (760 mm Hg)
Melting Pt: 295…297 °C
Storage Temperature: Ambient
MDL Number: MFCD00006318
CAS Number: 5345-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR