2-Aminonicotinamide 95%

Supplier: Ambeed
Warning

Synonyms: 2-Aminonicotinamide, 2-Aminopyridine-3-carboxamide

A509960-10G A509960-1G A509960-5G
103828-380EA 13 USD
103828-380 103839-480 77292-180 77307-652 77307-654
2-Aminonicotinamide 95%
2-Aminonicotinamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O
MW: 137.142 g/mol
Boiling Pt: 328.2 °C (760 mm Hg)
Melting Pt: 194…200 °C
Flash Pt: 152.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00128873
CAS Number: 13438-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR