2-Amino-3-formylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-pyridinecarboxaldehyde, 2-Aminonicotinaldehyde, 2-Aminopyridine-3-carboxaldehyde

A140988-100G A140988-10G A140988-500G A140988-1G A140988-25G A140988-5G
103802-930EA 9 USD
103802-930 77109-806 77109-808 77109-810 77316-054 77316-056 77316-058
2-Amino-3-formylpyridine 98%
2-Amino-3-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Melting Pt: 98…102 °C
Flash Pt: 129.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01830382
CAS Number: 7521-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR