2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 97%

Supplier: Ambeed
Warning

A357490-1G A357490-5G A357490-250MG
103826-194EA 7 USD
103826-194 103837-198 76840-352 76840-354 77263-956
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 97%
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClN₃OS
MW: 267.73 g/mol
Boiling Pt: 362 °C (760 mm Hg)
Melting Pt: 216…220 °C
Flash Pt: 172.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10000630
CAS Number: 302964-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR