Aminomalononitrile-p-toluenesulfonate, (max. 6.5% H₂O) 95% (contains < 6.5% H₂O)

Supplier: Ambeed
Warning

A138782-100G A138782-10G A138782-250MG A138782-1G
103802-666EA 6 USD
103802-666 76688-926 76688-928 76688-930 77315-728
Aminomalononitrile-p-toluenesulfonate, (max. 6.5% H₂O) 95% (contains < 6.5% H₂O)
Aminomalononitrile-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₃H₃N₃
Boiling Pt: 484 °C (760 mm Hg)
Melting Pt: 174 °C
Flash Pt: 246.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00012848
CAS Number: 5098-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR