2-Aminoisophthalic acid 98%

Supplier: Ambeed
Warning

A309780-1G A309780-5G A309780-100G A309780-10G A309780-250MG
103814-600EA 6 USD
103814-600 103835-782 76838-554 76838-556 77239-406 77239-408 77239-410
2-Aminoisophthalic acid 98%
2-Aminoisophthalic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
Boiling Pt: 429.7 °C (760 mm Hg)
Melting Pt: 312…315 °C
Flash Pt: 213.685 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00817808
CAS Number: 39622-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR