DL-α-Aminoadipic acid 97%

Supplier: Ambeed
Warning

Synonyms: DL-2-Aminoadipic acid, (RS)-2-Aminohexanedioic acid, H-DL-Homoglu-OH, 2-Aminohexanedioic acid, H-DL-Aad-OH

A112717-100G A112717-10G A112717-250MG A112717-25G A112717-1G A112717-5G
103798-084EA 20 USD
103798-084 103823-924 77160-832 77160-834 77160-836 77160-838 77297-202 77297-204
DL-α-Aminoadipic acid 97%
DL-α-Aminoadipic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₄
Melting Pt: 196…198 °C
Flash Pt: 173.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063119
CAS Number: 542-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR