2-Aminobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: o-Aminobenzophenone

A136592-100G A136592-10G A136592-500G A136592-1000G A136592-25G A136592-5G
103808-538EA 226 USD
103808-538 77109-576 77109-578 77109-580 77315-238 77315-240 77315-242
2-Aminobenzophenone 98%
2-Aminobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 383.7 °C (760 mm Hg)
Melting Pt: 103…107 °C
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00007713
CAS Number: 2835-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR