2-Aminobenzene-1,4-diol hydrochloride 95%

Supplier: Ambeed
Warning

A392688-100MG A392688-250MG A392688-1G
103826-908EA 494 USD
103826-908 103837-946 76817-790 76817-792 77307-018
2-Aminobenzene-1,4-diol hydrochloride 95%
2-Aminobenzene-1,4-diol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈ClNO₂
MW: 161.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13659348
CAS Number: 32190-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR