2-Aminobenzaldehyde 98%

Supplier: Ambeed
Warning

A104294-100G A104294-10G A104294-250MG A104294-25G A104294-1G A104294-5G
103796-680EA 6 USD
103796-680 103822-390 77257-388 77257-390 77257-392 77257-394 77283-196 77309-260
2-Aminobenzaldehyde 98%
2-Aminobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 258.7 °C (760 mm Hg)
Melting Pt: 38 °C
Flash Pt: 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00007709
CAS Number: 529-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR